Skip to Content
Merck
All Photos(1)

Key Documents

213837

Sigma-Aldrich

1-Piperidinecarbonitrile

99%

Synonym(s):

1-Cyanopiperidine, NSC 26965, Pentamethylenecyanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$162.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$162.00

About This Item

Empirical Formula (Hill Notation):
C6H10N2
CAS Number:
Molecular Weight:
110.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.47 (lit.)

bp

102 °C/10 mmHg (lit.)

density

0.951 g/mL at 25 °C (lit.)

SMILES string

N#CN1CCCCC1

InChI

1S/C6H10N2/c7-6-8-4-2-1-3-5-8/h1-5H2

InChI key

NVPICXQHSYQKGM-UHFFFAOYSA-N

General description

1-Piperidinecarbonitrile in tetrahydrofuran (THF) reacts with a solution of piperidyl lithium in THF at 0°C to yield lithium[1,1,3,3-bis(pentamethylene)guanidinate][1]. It participates in thermal decomposition of the 2H-azaphosphirene metal complexes to yield the Δ3-1,3,2-oxazaphospholene complexes and the Δ3-1,3,2-thiazaphospholene complexes[2].

Application

1-Piperidinecarbonitrile was used in the preparation of 1-selenocarbamoylpiperidine[3].
Reactant for:
Cycloaddition reactions
Ring-expansion reactions of tungsten complexes
N-CN bond cleavage
Retro-Diels-Alder reactions
Regioselective electrochemical α-methoxylation
Synthesis of de-O-chloroacetylation reagents

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel de-O-chloroacetylation reagent: 1-seleonocarbamoylpiperidine.
Sogabe S, et al.
Tetrahedron Letters, 47(37), 6603-6606 (2006)
Reactions of transiently formed nitrilium phosphanylide chromium, molybdenum, and tungsten complexes with heterocumulenes.
Streubel R and Neumann C.
Heteroatom Chem., 13(1), 72-76 (2002)
Winfried P Kretschmer et al.
Chemical communications (Cambridge, England), (6)(6), 608-609 (2002-07-18)
The titanium complex Cp[1,3-(2',6'-Me2C6H3)2(CH2N)2C=N]Ti(CH2Ph)2, with a monoanionic eta 1-iminoimidazolidide ancillary ligand, is shown to be a highly efficient catalyst for olefin polymerisation when activated with the Lewis acid B(C6F5)3.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service