Skip to Content
Merck
All Photos(5)

Documents

213217

Sigma-Aldrich

Lithium amide

greener alternative

powder, 95%

Synonym(s):

Aminolithium, Lithamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNH2
CAS Number:
Molecular Weight:
22.96
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

380-400 °C

density

1.178 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li].CN

InChI

1S/Li.H2N/h;1H2/q+1;-1

InChI key

AFRJJFRNGGLMDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium amide (LiNH2) is an amide of an alkali metal with lithium atoms having larger positive charges than hydrogen atoms. It is a moisture-sensitive molecule that converts to LiOH and NH3 on reaction with H2O. LiNH2 is majorly used in hydrogen storage applications.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Reagent for synthesis of:
Antiinflamatory and preoresolving protectin D1
Chemotype dipeptidyl peptidase IV inhibitors
Sterically congested triarylamines
Dyes displaying large Stokes shifts
GM1 ganglioside derivatives

Reagent for cross-coupling of aryl chlorides and amines

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. Ogawa and Y. Kobayashi,
Tetrahedron Letters, 52, 3001-3004 (2011)
G. Bordeau, et al.,
Tetrahedron Letters, 51, 4429-4432 (2010)
Rylan J Lundgren et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(6), 1983-1991 (2009-12-22)
The syntheses of 2-(di-tert-butylphosphino)-N,N-dimethylaniline (L1, 71%) and 2-(di-1-adamantylphosphino)-N,N-dimethylaniline (L2, 74 %), and their application in Buchwald-Hartwig amination, are reported. In combination with [Pd(allyl)Cl](2) or [Pd(cinnamyl)Cl](2), these structurally simple and air-stable P,N ligands enable the cross-coupling of aryl and heteroaryl chlorides
R. Kuwano, et al.,
Synlett, 12, 1819-1824 (2010)
Yukihiro Nishio et al.
Bioorganic & medicinal chemistry letters, 20(24), 7246-7249 (2010-11-16)
Structures containing the (R)-3-amino-3-methyl piperidine unit as a new pharmacophore moiety have been shown to possess moderate inhibitory activity for DPP-4 with good pharmacokinetics profile. One of these compounds was found to have good oral bioavailability and PK/PD profile in

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service