Skip to Content
Merck
All Photos(2)

Documents

213047

Sigma-Aldrich

2-(4-Chlorobenzoyl)benzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COC6H4CO2H
CAS Number:
Molecular Weight:
260.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

149-150 °C (lit.)

SMILES string

OC(=O)c1ccccc1C(=O)c2ccc(Cl)cc2

InChI

1S/C14H9ClO3/c15-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)14(17)18/h1-8H,(H,17,18)

InChI key

YWECCEXWKFHHQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(4-Chlorobenzoyl)benzoic acid forms complexes with europium and terbium having photoluminescence properties.

Application

2-(4-Chlorobenzoyl)benzoic acid was used in the preparation of bisphthalazinone monomers, required for the synthesis of phthalazinone containing poly(arylene ether)s, poly(arylene thioether)s and poly(arylene sulfone)s.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>572.0 °F

Flash Point(C)

> 300 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of Phthalazinone containing Poly (arylene ether) s, Poly (arylene thioether) s, and Poly (arylene sulfone) s via a novel NC coupling reaction.
Wang SJ, et al.
Macromolecules, 37(1), 60-65 (2004)
Zhao-Ye Li et al.
Guang pu xue yu guang pu fen xi = Guang pu, 29(11), 2904-2908 (2010-01-28)
Eight complexes of europium and terbium were synthesized, with 2-(4-chloro-benzoyl)benzoic acid as an essential ligand and other three distinct ligands, including 1, 10-phenanthroline, triphenylphosphine oxide and diphenyl sulfoxide, as selective ligands. They were Eu(CBBA)3 (H2O)4, Eu(CBBA)3 (Phen) (H2O)6, Eu(CBBA)3 (TPPO)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service