20910
Cadmium iodide
purum p.a., ≥99.0% (T)
Synonym(s):
Cadmium diiodide
Select a Size
About This Item
Recommended Products
grade
purum p.a.
Assay
≥99.0% (T)
form
powder
bp
787 °C (lit.)
mp
388 °C (lit.)
density
5.67 g/mL at 25 °C (lit.)
anion traces
bromide, chloride (as Cl-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
Ca: ≤100 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg
SMILES string
I[Cd]I
InChI
1S/Cd.2HI/h;2*1H/q+2;;/p-2
InChI key
OKIIEJOIXGHUKX-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | 464678 | 590703 | 202134 |
---|---|---|---|
assay ≥99.0% (T) | assay 97% | assay 99% | assay 99.9% trace metals basis |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
grade purum p.a. | grade - | grade - | grade - |
cation traces Ca: ≤100 mg/kg, Cu: ≤50 mg/kg, K: ≤100 mg/kg, Ni: ≤50 mg/kg, Zn: ≤50 mg/kg, Co: ≤50 mg/kg, Na: ≤100 mg/kg, Fe: ≤50 mg/kg, Pb: ≤50 mg/kg | cation traces - | cation traces - | cation traces - |
anion traces bromide, chloride (as Cl-): ≤200 mg/kg, sulfate (SO42-): ≤50 mg/kg | anion traces - | anion traces - | anion traces - |
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service