Skip to Content
Merck
All Photos(1)

Key Documents

208698

Sigma-Aldrich

3-Pentyn-1-ol

97%

Synonym(s):

1-Hydroxy-3-pentyne

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$144.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$144.00

About This Item

Linear Formula:
CH3C≡CCH2CH2OH
CAS Number:
Molecular Weight:
84.12
Beilstein:
1735967
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

154-157 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC#CCCO

InChI

1S/C5H8O/c1-2-3-4-5-6/h6H,4-5H2,1H3

InChI key

IDYNOORNKYEHHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Inhibiting effect of 3-pentyn-1-ol on the corrosion of iron in 1M HCl has been investigated[1]. Reaction of 3-pentyn-1-ol with Fe3(CO)12 in CH3OH/KOH solution yields closed trinuclear hydridic complex (μ-H)Fe3(CO)933-[H3CCC(CH2)(CH2)CO])[2].

Application

3-Pentyn-1-ol was used in the synthesis of 4,4-bis(1-methyl-1H-indol-3-yl)pentan-1-ol[3].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibitive properties, adsorption and surface study of butyn-1-ol and pentyn-1-ol alcohols as corrosion inhibitors for iron in HCl.
Babic-Samardzija K, et al.
Journal of Materials Chemistry, 15(19), 1908-1916 (2005)
Synthesis of bis (indolyl) alkanes by a site-selective gold-catalyzed addition of indoles to butynol derivatives.
Barluenga J, et al.
Journal of Organometallic Chemistry, 694(4), 546-550 (2009)
Reactions of Fe3(CO)12 with 2-methyl-3-butyn-1-ol and 3-pentyn-1-ol under basic methanolic conditions.
Bertolotti F, et al.
Journal of Organometallic Chemistry, 693(16), 2673-2678 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service