Skip to Content
Merck
All Photos(1)

Key Documents

208434

Sigma-Aldrich

Manganese(II) bromide tetrahydrate

98%

Synonym(s):

Manganese dibromide tetrahydrate, Manganese(2+) dibromide tetrahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$102.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$102.00

About This Item

Linear Formula:
MnBr2 · 4H2O
CAS Number:
Molecular Weight:
286.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

98%

form

crystalline

SMILES string

O.O.O.O.Br[Mn]Br

InChI

1S/2BrH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

HHDPJRSXPOGIOP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Dufour et al.
ACS nano, 13(5), 5326-5334 (2019-04-13)
Zinc blende II-VI semiconductor nanoplatelets (NPLs) are defined at the atomic scale along the thickness of the nanoparticle and are initially capped with carboxylates on the top and bottom [001] facets. These ligands are exchanged on CdSe NPLs with halides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service