Skip to Content
Merck
All Photos(1)

Key Documents

206253

Sigma-Aldrich

Ammonium hexachlororhodate(III)

reagent grade

Synonym(s):

Rhodium(III)-ammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$715.00

$715.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$715.00

About This Item

Linear Formula:
(NH4)3RhCl6
CAS Number:
Molecular Weight:
369.74
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
reagent grade
form:
powder
solubility:
H2O: slightly soluble(lit.)

$715.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

reagent grade

form

powder

solubility

H2O: slightly soluble(lit.)

SMILES string

[NH4+].[NH4+].[NH4+].Cl[Rh-3](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.3H3N.Rh/h6*1H;3*1H3;/q;;;;;;;;;+3/p-3

InChI key

OJNYBYILSUWBHM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lenka Svecova et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-12)
In this work, the aqueous speciation of Rh(III) in chloride medium was investigated by UV-vis spectroscopy for ligand to metal ratios R = (Cl-)/(Rh) ranging from 300 to 5000, at fixed Rh concentration (2.4 × 10-3 M). Under the chemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service