Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 g | Check Cart for Availability | $150.00 |
About This Item
Linear Formula:
S(CH2CH2CH2OH)2
CAS Number:
Molecular Weight:
150.24
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
EC Number:
234-202-7
MDL number:
Assay:
98%
Form:
liquid
Quality Segment
assay
98%
form
liquid
refractive index
n
bp
140-142 °C/0.5 mmHg (lit.)
density
1.092 g/mL at 25 °C (lit.)
functional group
hydroxyl, thioether
SMILES string
OCCCSCCCO
InChI
1S/C6H14O2S/c7-3-1-5-9-6-2-4-8/h7-8H,1-6H2
InChI key
QRQVZZMTKYXEKC-UHFFFAOYSA-N
Application
3,3′-Thiodipropanol was used in the determination of thiodiglycol in groundwater by a highly sensitive analytical method[1].
1 of 1
This Item | |||
|---|---|---|---|
| assay 98% | assay 96% | assay 98% | assay 98% |
| form liquid | form - | form - | form - |
| bp 140-142 °C/0.5 mmHg (lit.) | bp - | bp - | bp - |
| refractive index n | refractive index - | refractive index - | refractive index - |
| density 1.092 g/mL at 25 °C (lit.) | density - | density - | density - |
| functional group hydroxyl, thioether | functional group - | functional group - | functional group - |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



