Skip to Content
Merck
All Photos(1)

Key Documents

20425

Sigma-Aldrich

tert-Butoxy bis(dimethylamino)methane

technical

Synonym(s):

Bredereck’s reagent

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$393.00
50 ML
$1,750.00

$393.00


Estimated to ship onMay 12, 2025


Request a Bulk Order

Select a Size

Change View
10 ML
$393.00
50 ML
$1,750.00

About This Item

Linear Formula:
(CH3)3COCH[N(CH3)2]2
CAS Number:
Molecular Weight:
174.28
Beilstein:
1901973
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$393.00


Estimated to ship onMay 12, 2025


Request a Bulk Order

grade

technical

Quality Level

refractive index

n20/D 1.422 (lit.)
n20/D 1.422

bp

50-55 °C/15 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

functional group

amine
ether

SMILES string

CN(C)C(OC(C)(C)C)N(C)C

InChI

1S/C9H22N2O/c1-9(2,3)12-8(10(4)5)11(6)7/h8H,1-7H3

InChI key

HXRAMSFGUAOAJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reactant for:
  • Preparation of pyrroloquinazolines as photochemotherapeutic agents
  • Enanatioslective formal synthesis of the Cinchona alkaloid quinine via stereoselective intermolecular radical addition
  • Synthesis of camptothecin via intramolecular isomuenchnone cycloaddition reaction
  • Aminomethylenation reactions

Analysis Note

mixture of tert-Butoxy bis(dimethylamino)methane, tris(dimethylamino)methane and N,N-Dimethylformamide di-tert-butyl acetal

Other Notes

Review[1]; Mild condensation reactions[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Haefliger et al.
Tetrahedron Letters, 25, 285-285 (1984)
R.F. Abdulla et al.
Tetrahedron, 35, 1675-1675 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service