Skip to Content
Merck
All Photos(3)

Key Documents

201634

Sigma-Aldrich

6-Aminofluorescein

Synonym(s):

Fluoresceinamine isomer II

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$518.00
1 G
$1,580.00

$518.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$518.00
1 G
$1,580.00

About This Item

Empirical Formula (Hill Notation):
C20H13NO5
CAS Number:
Molecular Weight:
347.32
Beilstein:
51708
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$518.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

285 °C (dec.) (lit.)

solubility

methanol: 1 mg/mL, clear, yellow to very dark yellow-orange

λmax

495 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccc2C(=O)OC3(c4ccc(O)cc4Oc5cc(O)ccc35)c2c1

InChI

1S/C20H13NO5/c21-10-1-4-13-16(7-10)20(26-19(13)24)14-5-2-11(22)8-17(14)25-18-9-12(23)3-6-15(18)20/h1-9,22-23H,21H2

InChI key

YOAWSYSKQHLFPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Aminofluorescein has been demonstrated to be useful fluorescent labeling reagent for fullerene-based liposome nanostructures termed ‘buckysomes′. 6-aminofluorescein dissolved in 0.1 N NaOH can be quantified from a maximum absorbance at 490 nm. Decarboxylated 6-aminofluorescein dissolved 6 N HCl (prepare by constant boiling at 110° C for 24 h) can be quantified from a maximum absorbance at 454 nm. Aldehyde groups are formed by oxidateive damage to proteins, which can be detected by conjugation to decarboxylated 6-aminofluorescein.
6-Aminofluorescein has been used to determine its ability to potentiate the antiviral activity of poly r (A-U).[1]

Other Notes

Note: Do not confuse with fluorescamine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self assembly of amphiphilic C60 fullerene derivatives into nanoscale supramolecular structures
Partha R
Journal of Nanobiotechnology (2007)
I Climent et al.
Analytical biochemistry, 182(2), 226-232 (1989-11-01)
Oxidative modification of proteins is implicated in a number of physiologic and pathologic processes. Metal-catalyzed oxidative modification usually causes inactivation of enzymes and the appearance of carbonyl groups in amino acid side chains of the protein. We describe use of
W.E.G. Muller and H.C. Schroder
Biological Response Modifiers ? Interferons, Double-Stranded RNA and 2?,5?-Oligoadenylates (2012)
K C Ingham et al.
Biochimica et biophysica acta, 670(2), 181-189 (1981-09-29)
A method for covalent attachment of a fluorescent molecule to the carbohydrate moieties of glycoproteins is described. The glycoproteins were oxidized with periodate under mild conditions selective for sialic acid (Van Lenten, L. and Ashwell, G. (1971) J. Biol. Chem.
Miju Kim et al.
ACS nano, 9(8), 8269-8278 (2015-07-15)
Polyelectrolyte microcapsules represent versatile stimuli-responsive structures that enable the encapsulation, protection, and release of active agents. Their conventional preparation methods, however, tend to be time-consuming, yield low encapsulation efficiency, and seldom allow for the dual incorporation of hydrophilic and hydrophobic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service