Skip to Content
Merck
All Photos(1)

Key Documents

199915

Sigma-Aldrich

1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine

97%

Synonym(s):

MPTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N
CAS Number:
Molecular Weight:
173.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

form

solid

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

bp

128-132 °C/12 mmHg (lit.)

functional group

phenyl

SMILES string

CN1CCC(=CC1)c2ccccc2

InChI

1S/C12H15N/c1-13-9-7-12(8-10-13)11-5-3-2-4-6-11/h2-7H,8-10H2,1H3

InChI key

PLRACCBDVIHHLZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dopaminergic neurotoxin.

Warning

Reportedly causes a severe and irreversible Parkinsonism condition in humans and monkeys. New Engl. J. Med. 1983, 309, 310. Science 1983, 219, 979. ibid. 1984, 224, 1083, 1451. ibid. 1984, 225, 529. J. Med. Chem. 1985, 28, 997.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideaki Matsui et al.
Journal of neural transmission (Vienna, Austria : 1996), 125(1), 25-33 (2017-08-05)
Parkinson's disease is a neurodegenerative disorder that involves movement discloses, degeneration of dopaminergic neurons, and presence of cytoplasmic inclusion bodies. Various animal models have been developed and small fish including zebrafish and medaka fish have recently been employed as a
Gejuan Zhang et al.
Life sciences, 232, 116600-116600 (2019-06-30)
Neuroinflammation is one of the significant neuropathological conditions in Parkinson's disease (PD) which is due to microglial and astrocytes activation leads to progressive dopaminergic neuronal loss. To date, Current PD drugs offers only symptomatic relief with adverse effects and lack
Yuanyuan Liu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 40(1-2), 18-26 (2016-11-15)
Development of effective therapeutic drugs for Parkinson's disease is in great need. During the progression of Parkinson's disease, Rho-associated protein kinase 2 (ROCK2) is activated to promote neurodegeneration. Hydrogen sulfide (H2S) has a neuroprotective effect during the neural injury of
David I Finkelstein et al.
Acta neuropathologica communications, 5(1), 53-53 (2017-07-01)
Elevated iron in the SNpc may play a key role in Parkinson's disease (PD) neurodegeneration since drug candidates with high iron affinity rescue PD animal models, and one candidate, deferirpone, has shown efficacy recently in a phase two clinical trial.
Luisa Quinti et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(23), E4676-E4685 (2017-05-24)
The activity of the transcription factor nuclear factor-erythroid 2 p45-derived factor 2 (NRF2) is orchestrated and amplified through enhanced transcription of antioxidant and antiinflammatory target genes. The present study has characterized a triazole-containing inducer of NRF2 and elucidated the mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service