Skip to Content
Merck
All Photos(1)

Key Documents

199192

Sigma-Aldrich

3-(Trifluoromethyl)phenylacetonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$65.10

$65.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$65.10

About This Item

Linear Formula:
CF3C6H4CH2CN
CAS Number:
Molecular Weight:
185.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$65.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.4565 (lit.)

bp

92-93 °C/4 mmHg (lit.)

density

1.187 g/mL at 25 °C (lit.)

functional group

fluoro
nitrile

SMILES string

FC(F)(F)c1cccc(CC#N)c1

InChI

1S/C9H6F3N/c10-9(11,12)8-3-1-2-7(6-8)4-5-13/h1-3,6H,4H2

InChI key

JOIYKSLWXLFGGR-UHFFFAOYSA-N

Application

3-(Trifluoromethyl)phenylacetonitrile may be used in chemical synthesis studies.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service