Skip to Content
Merck
All Photos(3)

Key Documents

198102

Sigma-Aldrich

Sudan IV

Certified Biological Stain Commission,Dye content ≥80 %

Synonym(s):

Biebrich scarlet R fat soluble, Fat Ponceau R or 4, Lipid Crimson, Oil Red IV, Scarlet Red Scharlach, Solvent Red 24, Sudan R or BB

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$162.00
100 G
$341.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$162.00
100 G
$341.00

About This Item

Empirical Formula (Hill Notation):
C24H20N4O
CAS Number:
Molecular Weight:
380.44
Colour Index Number:
26105
Beilstein:
709018
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

composition

Certified Biological Stain Commission,Dye content, ≥80%

technique(s)

microbe id | staining: suitable

mp

199 °C (dec.) (lit.)

λmax

357 nm (2nd)
520 nm

ε (extinction coefficient)

≥12000 at 356-360 nm in toluene at 0.008 g/L
≥26000 at 519-523 nm in toluene at 0.008 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccccc1\N=N\c2ccc(\N=N\c3c(O)ccc4ccccc34)c(C)c2

InChI

1S/C24H20N4O/c1-16-7-3-6-10-21(16)26-25-19-12-13-22(17(2)15-19)27-28-24-20-9-5-4-8-18(20)11-14-23(24)29/h3-15,29H,1-2H3/b26-25+,28-27+

InChI key

RCTGMCJBQGBLKT-PAMTUDGESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sudan IV is a hydrophobic azo dye.[1][2] It stains triglycerides and imparts an intense red color. The dye does not stain lipid droplets or phospholipids.[3]

Application

Sudan IV has been used to in en-face staining of aorta, stains the lipid rich plaques.[4] It has also been used to stain lipids atherosclerotic lesions of mice.[4][5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The
Histophatologic Techniques (2006)
In Situ Remediation of Chlorinated Solvent Plumes (2010)
Aorta atherosclerosis lesion analysis in hyperlipidemic mice
Mohanta S, et al.
Bio-protocol, 6(11), 77-84 (2016)
The induction of cytochrome P450 1A1 by Sudan dyes
Ahmed R, et al.
Journal of Biochemical and Molecular Toxicology, 22(2), 77-84 (2008)
Discovery of a potent HMG-CoA reductase degrader that eliminates statin-induced reductase accumulation and lowers cholesterol
Jiang SY, et al.
Nature Communications, 9(1), 5138-5138 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service