Skip to Content
Merck
All Photos(1)

Documents

197416

Sigma-Aldrich

3-(4-Hydroxyphenyl)-1-propanol

99%

Synonym(s):

1-(4-Hydroxyphenyl)-3-propanol, 3-(4-Hydroxyphenyl)propanol, 3-(p-Hydroxyphenyl)-1-propanol, 3-(p-Hydroxyphenyl)propyl alcohol, 4-(3-Hydroxypropyl)phenol, 4-Hydroxybenzenepropanol, Dihydro-p-coumaryl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4(CH2)3OH
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

51-54 °C (lit.)

SMILES string

OCCCc1ccc(O)cc1

InChI

1S/C9H12O2/c10-7-1-2-8-3-5-9(11)6-4-8/h3-6,10-11H,1-2,7H2

InChI key

NJCVPQRHRKYSAZ-UHFFFAOYSA-N

Application

3-(4-Hydroxyphenyl)-1-propanol was used in the synthesis of (−)-centrolobine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yangxin Wang et al.
Frontiers in bioengineering and biotechnology, 8, 714-714 (2020-08-01)
In situ immobilization of enzyme into metal-organic frameworks (MOFs) is performed through a one-step and facile method. Candida antarctica lipase B (CalB) is directly embedded in zeolitic imidazolate framework (ZIF)-8 by simply mixing an aqueous solution of 2-methylimidazole and zinc
Anna Boronat et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-06)
Beer is a fermented beverage with beneficial phenolic compounds and is widely consumed worldwide. The current study aimed to describe the content of three families of phenolic compounds with relevant biological activities: prenylated flavonoids (from hops), simple phenolic alcohols (from
Rosa Cert et al.
Food & function, 6(6), 1999-2007 (2015-05-29)
The antioxidant activity of ethyl and octyl hydroxytyrosyl ethers toward lipids was determined using the Rancimat and open cup methods at high temperatures and 50 °C, respectively. The effect of the unsaturation of the matrix was evaluated using sunflower, soya
Two successive one-pot reactions leading to the expeditious synthesis of (−)-centrolobine.
Tetrahedron Letters, 45(35), 6603-6605 (2004)
Georgios Koutrotsios et al.
Journal of agricultural and food chemistry, 66(24), 5971-5983 (2018-06-06)
Pleurotus ostreatus, P. eryngii, and P. nebrodensis were cultivated on nonconventional substrates containing grape marc (GMC) or olive mill byproducts (OMB); wheat straw (WHS) served as control. GMC-based media demonstrated equal/better mushroom productivity than WHS for P. eryngii and P.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service