Skip to Content
Merck
All Photos(2)

Key Documents

195596

Sigma-Aldrich

sec-Butyllithium solution

sec-Butyllithium solution
1 of 3 reviewers received a sample product or took part in a promotion

1.4 M in cyclohexane

Synonym(s):

Lithium-2-butanide, s-BuLi

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
3587206
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

concentration

1.4 M in cyclohexane

density

0.769 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]C(C)CC

InChI

1S/C4H9.Li/c1-3-4-2;/h3H,4H2,1-2H3;

InChI key

VATDYQWILMGLEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

sec-Butyllithium solution (1.4M in cyclohexane) has been used in the multi-step synthesis of 5-methyl-5,6-dihydrothymidine (5-MDHT) from thymidine.[1] It has also been used in the copper-catalyzed asymmetric allylic alkylation (AAA) of allyl bromides, chlorides, and ethers in the presence of a chiral ligand.[2]

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng Chen et al.
Organic & biomolecular chemistry, 15(25), 5364-5372 (2017-06-16)
A series of 6,6-dihalo-2-azabicyclo[3.1.0]hexane and 7,7-dihalo-2-azabicyclo[4.1.0]heptane compounds were prepared by the reaction of dihalocarbene species with N-Boc-2,3-dihydro-1H-pyrroles or -1,2,3,4-tetrahydropyridines. Monochloro substrates were synthesised as well, using a chlorine-to-lithium exchange reaction. The behaviour of several aldehydes and ketones under reductive amination
Leixing Chen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(68), 18012-18019 (2018-11-15)
Although living polymerization methods are widely applicable to organic monomers, their application to inorganic monomers is rare. For the first time, we show that the living poly(methylenephosphine) (PMPn- ) anion can function as a macroinitiator for olefins. Specifically, the phosphaalkene
Dimitrios Moschovas et al.
Nanomaterials (Basel, Switzerland), 10(8) (2020-08-06)
The synthesis, molecular and morphological characterization of a 3-miktoarm star terpolymer of polystyrene (PS, M¯n = 61.0 kg/mol), polybutadiene (PB, M¯n = 38.2 kg/mol) and polyisoprene (PI, M¯n = 29.2 kg/mol), corresponding to volume fractions (φ) of 0.46, 0.31 and
Jason V Chari et al.
The Journal of organic chemistry, 84(6), 3652-3655 (2019-03-07)
Silyl triflate precursors to cyclic alkynes and allenes serve as valuable synthetic building blocks. We report a concise and scalable synthetic approach to prepare the silyl triflate precursors to cyclohexyne and 1,2-cyclohexadiene. The strategy involves a retro-Brook rearrangement of an
Lukasz Otulakowski et al.
Polymers, 12(1) (2020-01-17)
In this work, the self-assembly of a series of amphiphilic polystyrene-b-polyglycidol (PS-b-PGL) diblock copolymers in dioxane and dioxane/water mixtures is presented. The PS-b-PGL have an average degree of polymerization (DP) of PS block equal to 29 units and varied degrees

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1 of 3 reviewers received a sample product or took part in a promotion
1–2 of 3 Reviews  

Active Filters

  1. South West, UK
    • Review 1
    • Votes 7
    2 out of 5 stars.

    Titrate your s-BuLi before use

    s-BuLi when titrated came to about 0.73 M not the 1.4 M stated on the bottle

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for taking the time to leave a review! sec-Butyllithium is very sensitive to air, moisture and temperature and should be handled under inert atmosphere. During storage a white precipitate of lithium hydride may form as it slowly decomposes, which is part of the characteristics of this chemical resulting in a limited shelf life. We would love to learn more about your experience and see if there is anything more we can do to help with this issue. When you have a moment, please contact us by visiting https://www.sigmaaldrich.com/support/customer-support and submit a Product Technical Inquiry. We look forward to hearing from you soon!

  2. South West
    • Review 1
    • Votes 6
    1 out of 5 stars.

    Highly turbid, with sediment that doesn't settle

    Highly turbid, with sediment that doesn't settle, giving inconsistent results. Titre is less than half than marketed.

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for taking the time to leave a review! We are sorry to hear that your experience did not meet expectations. We would like to learn more about your experience to see if there is anything we can do to help troubleshoot this issue. When you have a moment, please contact us by visiting https://www.sigmaaldrich.com/support/customer-support and submit a Report Product Issue ticket. We look forward to hearing from you soon!

  3. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service