Skip to Content
Merck
All Photos(1)

Key Documents

194719

Sigma-Aldrich

Vincamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O3
CAS Number:
Molecular Weight:
354.44
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]23/D +42.8°, c = 1 in pyridine

mp

232 °C (dec.) (lit.)

SMILES string

CC[C@@]12CCCN3CCc4c(C13)n(c5ccccc45)[C@](O)(C2)C(=O)OC

InChI

1S/C21H26N2O3/c1-3-20-10-6-11-22-12-9-15-14-7-4-5-8-16(14)23(17(15)18(20)22)21(25,13-20)19(24)26-2/h4-5,7-8,18,25H,3,6,9-13H2,1-2H3/t18-,20+,21+/m1/s1

InChI key

RXPRRQLKFXBCSJ-GIVPXCGWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Vincamine can be used as a starting material to synthesize:
  • Vincamine acid, which is employed as a precursor in the synthesis of vinpocetine by dehydration and esterification using sulfuric acid.[1]
  • Apovincamine using iron(III) perchlorate.[2]
  • (-)-Criocerine via one-step iodination reaction.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Adám Vas et al.
Medicinal research reviews, 25(6), 737-757 (2005-09-15)
The Apocynaceae plant family contains a great number of so called eburnamine-vincamine alkaloids. Quite a few of these alkaloids exert varied pharmacological activities on the cell multiplication, cardiovascular system, and brain functions. Many derivatives were also synthesized to find pharmacologically
K Cambon et al.
Neuroscience, 171(3), 840-851 (2010-10-07)
The Balb/c strain and the C57BL/6 strain show constitutive differences for tyrosine hydroxylase expression, and noradrenaline (NA) prefrontal transmission. Male mice of these strains also show striking differences in social interaction behaviors, with an increased aggressiveness for the Balb/c strain.
S Caccamese et al.
Journal of chromatography. A, 893(1), 47-54 (2000-10-24)
The four enantiomeric pairs of vincamine group alkaloids were separated by HPLC using Chiralpak AD as chiral stationary phase (CSP) and various n-hexane-2-propanol and n-hexane-ethanol mobile phases. (+)-cis-Vincamine, which is used in pharmaceutical preparations, is eluted much faster than its
Hong Yin et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(8-9), 802-805 (2011-02-15)
Vinca minor is a plant containing the alkaloid vincamine, which is used in the pharmaceutical industry as a cerebral stimulant and vasodilator. The objective of this study was to determine whether endophytic fungi isolated from V. minor produce vincamine. Primary
Mazák Károly et al.
Journal of chromatography. A, 996(1-2), 195-203 (2003-07-02)
A reversed-phase thin-layer chromatographic method was developed and applied to quantitate the lipophilicity of sparingly water-soluble eburnane alkaloids of therapeutic interest. Our method development included calibration, optimization and validation procedures, using also sets of auxiliary compounds. The log P(TLC) values

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service