Skip to Content
Merck
All Photos(1)

Documents

194492

Sigma-Aldrich

3-Ethoxyaniline

98%

Synonym(s):

m-Phenetidine, 3-Ethoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
971028
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.566 (lit.)

bp

248 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

SMILES string

CCOc1cccc(N)c1

InChI

1S/C8H11NO/c1-2-10-8-5-3-4-7(9)6-8/h3-6H,2,9H2,1H3

InChI key

WEZAHYDFZNTGKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Ethoxyaniline was used as starting reagent in the synthesis of 1,2,3,4-tetrahydroacridinylhydrazides.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Gemma et al.
Bioorganic & medicinal chemistry letters, 21(9), 2776-2779 (2010-10-01)
Among the enzymes involved in the life cycle of HCV, the non-structural protein NS3, with its double function of protease and NTPase/helicase, is essential for the virus replication. Exploiting our previous knowledge in the development of nucleotide-mimicking NS3 helicase (NS3h)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service