Skip to Content
Merck
All Photos(2)

Key Documents

194239

Sigma-Aldrich

Dimethyl isophthalate

ReagentPlus®, 99%

Synonym(s):

1,3-Di(methoxycarbonyl)benzene, 1,3-Dimethyl benzene-1,3-dicarboxylate, DMIP, Dimethy isophthalate, Dimethyl 1,3-benzenedicarboxylate, Dimethyl m-phthalate, Methyl 3-carbomethoxybenzoate, Methyl isophthalate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$61.20
500 G
$107.00
3 KG
$350.00

$61.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$61.20
500 G
$107.00
3 KG
$350.00

About This Item

Linear Formula:
C6H4-1,3-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1912251
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

6.8 (vs air)

Quality Level

vapor pressure

93.7 mmHg ( 208 °C)

product line

ReagentPlus®

Assay

99%

mp

64-68 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1cccc(c1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-4-3-5-8(6-7)10(12)14-2/h3-6H,1-2H3

InChI key

VNGOYPQMJFJDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl isophthalate is dimethyl phthalate ester based plasticizer[1].

Application

Dimethyl isophthalate was used in the synthesis of:
  • polybutylene isophthalate end-capped with N-ethyl-pentadecafluorooctanamide[2]
  • poly(ethylene terephthalate-co-isophthalate-co-5-tert-butylisophthalate) terpolyesters[3]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

343.4 °F - closed cup

Flash Point(C)

173 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(ethylene terephthalate) terpolyesters containing isophthalic and 5-tert-butylisophthalic units
Kint DPR, et al.
Journal of Polymer Science: Part A, General Papers, 41(1), 124-134 (2003)
Zhonggang Wang et al.
The journal of physical chemistry. B, 113(46), 15204-15211 (2009-10-22)
Polybutylene isophthalates (PBI) end-capped with N-ethyl-pentadecafluorooctanamide were synthesized from dimethyl isophthalate, 1,4-butanediol, and N-(2-hydroxyethyl)-perfluorooctanamide with fluorine monomer content from 0 to 5 mol %. The results demonstrated that there was an obvious gradient drop of fluorine concentration from the film
Zhu-Hua Luo et al.
Environmental science and pollution research international, 18(9), 1653-1660 (2011-06-01)
Dimethyl phthalate esters (DMPEs) are a group of plasticizers commonly detected in the environment with potential adverse human health impact. The degradation of DMPEs by fungal systems has been studied to a limited extent, particularly by yeasts. In this study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service