191914
Polyethylene, oxidized
average Mw ~3,600 by GPC, average Mn ~1,300 by GPC
About This Item
Recommended Products
mol wt
average Mn ~1,300 by GPC
average Mw ~3,600 by GPC
viscosity
225 cP(125 °C, Brookfield Thermosel)(lit.)
acid number
16 mg KOH/g
transition temp
softening point 104 °C (ring and ball)
density
0.93 g/mL at 25 °C
Looking for similar products? Visit Product Comparison Guide
Application
Physical properties
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service