Skip to Content
Merck
All Photos(3)

Key Documents

190942

Sigma-Aldrich

Poly(1,4-butylene terephthalate)

average Mv ~38,000, pellets

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$313.00

$313.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$313.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$313.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

pellets

Quality Level

mol wt

average Mv ~38,000

transition temp

Tm 227 °C
Tg 66 °C

density

1.31 g/mL at 25 °C (lit.)

SMILES string

OCCCCO.OC(=O)c1ccc(cc1)C(O)=O

InChI

1S/C10H10O4.C4H10O2/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2;5-3-1-2-4-6/h3-6H,1-2H3;5-6H,1-4H2

InChI key

DHQLHXKTPYFDCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Molded into auto parts, electrical components and domestic appliance assemblies.

Features and Benefits

Chemical resistance similar to PET. Low water absorption giving improved dimensional stability and electrical insulating properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denisse Esther Mallaupoma Camarena et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-20)
The development of products for topical applications requires analyses of their skin effects before they are destined for the market. At present, the ban on animal use in several tests makes the search for in vitro models (such as artificial
Masaki Watanabe et al.
Polymers, 12(1) (2020-01-16)
The purpose of this study was to evaluate the physical and chemical properties of engineering plastics processed using supercritical CO2. First, we prepared disk-shaped test pieces via a general molding process, which were plasticized using supercritical CO2 at temperatures lower

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service