Skip to Content
Merck
All Photos(1)

Key Documents

190675

Sigma-Aldrich

4-Methyl-5-thiazoleethanol

98%

Synonym(s):

5-(2-Hydroxyethyl)-4-methylthiazole, Sulfurol

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$51.60

$51.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$51.60

About This Item

Empirical Formula (Hill Notation):
C6H9NOS
CAS Number:
Molecular Weight:
143.21
Beilstein:
114249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$51.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.550 (lit.)

bp

135 °C/7 mmHg (lit.)

solubility

alcohol: soluble(lit.)
benzene: soluble(lit.)
chloroform: soluble(lit.)
diethyl ether/hexanes: soluble(lit.)
water: very soluble(lit.)

density

1.196 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

Cc1ncsc1CCO

InChI

1S/C6H9NOS/c1-5-6(2-3-8)9-4-7-5/h4,8H,2-3H2,1H3

InChI key

BKAWJIRCKVUVED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-5-thiazoleethanol reacts with 4-ethyloctanoyl chloride to yield 4-methyl-5-thiazoleethanol 4-ethyloctanoate, sulfur-containing flavor compound[1]. It was isolated from ether soluble alkaloidal fraction of Panax ginseng[2].

Application

4-Methyl-5-thiazoleethanol has been used in the synthesis of novel thiazolium halogenide ionic liquids[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses and Investigation of Properties of New Ionic Liquids Thiazolium Halogenide.
Wang QK, et al.
Advanced Materials Research, 332, 2036-2039 (2011)
A thiazole and two β-carboline constitutents from Panax ginseng.
Park JD, et al.
Archives of Pharmacal Research, 11(1), 52-55 (1988)
C Niederberger et al.
Gene, 171(1), 119-122 (1996-05-24)
Amiloride (Am) inhibits growth in the fission yeast Schizosaccharomyces pombe. We show that the toxic effect of this drug is relieved by low concentrations of thiamine (Th) and that the pyrimidine moiety of the Th molecule is responsible for growth
Yuping Liu et al.
Molecules (Basel, Switzerland), 15(8), 5104-5111 (2010-08-18)
Five sulfur-containing flavor compounds were synthesized for the first time by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. The synthesized compounds are 3-(methylthio)propyl 4-ethyloctanoate, 2-methyl-3-tetrahydro-furanthiol 4-ethyloctanoate, 4-methyl-5-thiazoleethanol 4-ethyloctanoate, 2-furan-methanethiol 4-ethyloctanoate and 2-methyl-3-furanthiol 4-ethyloctanoate. These five synthetic sulfur-containing
Biosynthesis of thiamin thiazole: determination of the regiochemistry of the S/O acyl shift by using 1,4-dideoxy-D-xylulose-5-phosphate.
Abhishek Chatterjee et al.
Angewandte Chemie (International ed. in English), 45(21), 3507-3510 (2006-04-21)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service