190543
Acrolein dimethyl acetal
98%
Synonym(s):
3,3-Dimethoxy-1-propene
About This Item
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.395 (lit.)
bp
89-90 °C (lit.)
density
0.862 g/mL at 25 °C (lit.)
functional group
acetal
ether
SMILES string
COC(OC)C=C
InChI
1S/C5H10O2/c1-4-5(6-2)7-3/h4-5H,1H2,2-3H3
InChI key
OBWGMYALGNDUNM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
26.6 °F - closed cup
Flash Point(C)
-3 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service