Skip to Content
Merck
All Photos(1)

Key Documents

190527

Sigma-Aldrich

(S)-(+)-Isoleucinol

97%

Synonym(s):

(2S,3S)-2-Amino-3-methyl-1-pentanol, L-Isoleucinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$72.70

$72.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$72.70

About This Item

Linear Formula:
C2H5CH(CH3)CH(NH2)CH2OH
CAS Number:
Molecular Weight:
117.19
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$72.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

optical activity

[α]20/D +4.0°, c = 1.6 in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.4589 (lit.)

bp

97 °C/14 mmHg (lit.)

mp

36-39 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC[C@H](C)[C@H](N)CO

InChI

1S/C6H15NO/c1-3-5(2)6(7)4-8/h5-6,8H,3-4,7H2,1-2H3/t5-,6+/m0/s1

InChI key

VTQHAQXFSHDMHT-NTSWFWBYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service