Skip to Content
Merck
All Photos(1)

Key Documents

188360

Sigma-Aldrich

(1R)-(−)-10-Camphorsulfonic acid ammonium salt

98%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$177.00

$177.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$177.00

About This Item

Empirical Formula (Hill Notation):
C10H19NO4S
CAS Number:
Molecular Weight:
249.33
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

$177.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

optical activity

[α]22/D −18.4°, c = 5.3 in H2O

mp

250 °C (dec.) (lit.)

functional group

ketone
sulfonic acid

SMILES string

N.CC1(C)[C@H]2CC[C@]1(CS(O)(=O)=O)C(=O)C2

InChI

1S/C10H16O4S.H3N/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14;/h7H,3-6H2,1-2H3,(H,12,13,14);1H3/t7-,10-;/m0./s1

InChI key

JTMZBRWRXFAITF-YUWZRIFDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1R)-(-)-10-Camphorsulfonic acid ammonium salt may be used as an ion-pair reagent to enhance the ability of supercritical carbon dioxide to extract polar compounds in supercritical fluid extraction (SFE) process.[1][2] It can also be used in the synthesis of (-)-quadrone.[3]

Application

Reactant involved in the synthesis of chiral anionic liquids for use as solvents and chiral shift reagents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion-pair-supercritical fluid extraction of clenbuterol from food samples.
Jimenez-Carmona MM, et al.
Journal of Chromatography A, 711(2), 269-276 (1995)
Total synthesis of natural quadrone.
Liu HJ and Llinas-Brunet M.
Canadian Journal of Chemistry, 66(3), 528-530 (1988)
Sub-and supercritical fluid extraction of trichloropyridinol from soil prior to immunoassay.
Jimenez-Carmona MM, et al.
Journal of Chromatography A, 785(1-2), 329-336 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service