Skip to Content
Merck
All Photos(1)

Key Documents

187402

Sigma-Aldrich

5-Chloro-2-nitrobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$337.00

$337.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$337.00

About This Item

Linear Formula:
ClC6H3(NO2)CH2OH
CAS Number:
Molecular Weight:
187.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$337.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

mp

78-79 °C (lit.)

functional group

chloro
hydroxyl
nitro

SMILES string

OCc1cc(Cl)ccc1[N+]([O-])=O

InChI

1S/C7H6ClNO3/c8-6-1-2-7(9(11)12)5(3-6)4-10/h1-3,10H,4H2

InChI key

ULYZTHQGJXPEFT-UHFFFAOYSA-N

Application

5-Chloro-2-nitrobenzyl alcohol was used in the synthesis of carbamates[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of carbamates from amines and alcohols under mild conditions.
D'Addona D and Bochet CG.
Tetrahedron Letters, 42(31), 5227-5229 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service