Skip to Content
Merck
All Photos(1)

Key Documents

187372

Sigma-Aldrich

4-Benzyloxybenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4CH2OH
CAS Number:
Molecular Weight:
214.26
Beilstein:
1877819
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

86-87 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

OCc1ccc(OCc2ccccc2)cc1

InChI

1S/C14H14O2/c15-10-12-6-8-14(9-7-12)16-11-13-4-2-1-3-5-13/h1-9,15H,10-11H2

InChI key

OEBIVOHKFYSBPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Benzyloxybenzyl alcohol was used in the synthesis of:
  • 13C-labelled derivatives of the isoflavonoid phytoestrogens, genistein, biochanin A, daidzein and formononetin[1]
  • benzyl4-(tert-butyldiphenylsiloxy-M1-carbamoyloxymethyl)phenylether[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica Tonolo et al.
Antioxidants (Basel, Switzerland), 9(2) (2020-02-06)
Due to their beneficial properties, fermented foods are considered important constituents of the human diet. They also contain bioactive peptides, health-promoting compounds studied for a wide range of effects. In this work, several antioxidant peptides extracted from fermented milk proteins
The synthesis of precisely structured polyurethanes. Part 2. Chain building methodology.
George MH, et al.
Journal of the Chemical Society. Perkin Transactions 1, 12, 1395-1401 (1996)
Synthesis of [4-13C]-Isoflavonoid Phytoestrogens.
Whalley JL, et al.
Tetrahedron, 56(3), 455-460 (2000)
Alexandre Murza et al.
Organic & biomolecular chemistry, 15(2), 449-458 (2016-12-08)
Apelin is the endogenous ligand for the G protein-coupled receptor APJ and exerts a key role in regulating cardiovascular functions. We report herein a novel series of macrocyclic analogues of apelin-13 in which the N- and C-terminal residues as well
Armine Avetisyan et al.
Neurochemistry international, 140, 104799-104799 (2020-08-14)
The receptor for advanced glycation end products (RAGE) is considered to contribute to the pathogenesis of Alzheimer's disease (AD), mediating amyloid beta (Aβ) accumulation, mitochondrial damage, and neuroinflammation. Previously, we have synthesized small peptides corresponding to the fragments (60-76) (P1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service