Skip to Content
Merck
All Photos(1)

Key Documents

187267

Sigma-Aldrich

4-Methyl-3-nitrobenzyl alcohol

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$40.30

$40.30


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$40.30

About This Item

Linear Formula:
CH3C6H3(NO2)CH2OH
CAS Number:
Molecular Weight:
167.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$40.30


Estimated to ship onMay 25, 2025


Request a Bulk Order

Assay

≥98%

form

solid

mp

39-43 °C (lit.)

functional group

hydroxyl
nitro

SMILES string

Cc1ccc(CO)cc1[N+]([O-])=O

InChI

1S/C8H9NO3/c1-6-2-3-7(5-10)4-8(6)9(11)12/h2-4,10H,5H2,1H3

InChI key

URCWIFSXDARYLY-UHFFFAOYSA-N

Application

4-Methyl-3-nitrobenzyl alcohol may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service