Skip to Content
Merck
All Photos(1)

Key Documents

18717

Sigma-Aldrich

(3R,6R)-3,6-Octanediol

≥98.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18O2
CAS Number:
Molecular Weight:
146.23
Beilstein:
4780881
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥98.0% (sum of enantiomers, GC)

optical purity

enantiomeric ratio: ≥99:1

SMILES string

CC[C@@H](O)CC[C@H](O)CC

Application

Used as a convenient starting material for the preparation of enantiomerically pure bis(phospholano)ethane ligands.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burk, M.J. et al.
Journal of the American Chemical Society, 115, 10125-10125 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service