Skip to Content
Merck
All Photos(1)

Key Documents

186228

Sigma-Aldrich

2′-Aminoacetophenone hydrochloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC6H4NH2 · HCl
CAS Number:
Molecular Weight:
171.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

285-289 °C (lit.)

SMILES string

Cl.CC(=O)c1ccccc1N

InChI

1S/C8H9NO.ClH/c1-6(10)7-4-2-3-5-8(7)9;/h2-5H,9H2,1H3;1H

InChI key

APTPPYQXBFFQHZ-UHFFFAOYSA-N

Application

2′-Aminoacetophenone hydrochloride was used in the synthesis of α-benzamidoacetophenone and indazoles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Preparation of Indazoles Via Metal Free Intramolecular Electrophilic Amination of 2-Aminophenyl Ketoximes.
M Counceller C, et al.
Organic Syntheses, 33-41 (2012)
Chloramphenicol1 (Chloromycetin). VI. A Synthetic Approach.
Long LM and Troutman HD.
Journal of the American Chemical Society, 71(7), 2469-2472 (1949)
Andrew P Worth et al.
Methods in molecular biology (Clifton, N.J.), 930, 125-162 (2012-10-23)
In this chapter, a range of computational tools for applying QSAR and grouping/read-across methods are described, and their integrated use in the computational assessment of genotoxicity is illustrated through the application of selected tools to two case-study compounds-2-amino-9H-pyrido[2,3-b]indole (AαC) and
R E Page et al.
Experientia, 44(3), 270-271 (1988-03-15)
o-Aminoacetophenone is a pheromone produced by virgin honeybee queens and released with feces. In small social groups, the pheromone repels and is used to terminate agonistic interactions between queens and workers.
Meenu Kesarwani et al.
PLoS pathogens, 7(8), e1002192-e1002192 (2011-08-11)
A significant number of environmental microorganisms can cause serious, even fatal, acute and chronic infections in humans. The severity and outcome of each type of infection depends on the expression of specific bacterial phenotypes controlled by complex regulatory networks that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service