184888
Naphthalene-bis(hexachlorocyclopentadiene) adduct
technical grade, 95%
Synonym(s):
DHA, NSC 122892
About This Item
grade
technical grade
Assay
95%
mp
208-210 °C (lit.)
SMILES string
ClC1=C(Cl)[C@@]2(Cl)C3C(C4C(c5ccccc35)[C@@]6(Cl)C(Cl)=C(Cl)[C@]4(Cl)C6(Cl)Cl)[C@]1(Cl)C2(Cl)Cl
InChI
1S/C20H8Cl12/c21-11-13(23)17(27)9-7(15(11,25)19(17,29)30)5-3-1-2-4-6(5)8-10(9)18(28)14(24)12(22)16(8,26)20(18,31)32/h1-4,7-10H/t7?,8?,9?,10?,15-,16+,17+,18-
InChI key
ULBZLTIPWBXWCY-QDCKJYKTSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Molecule studied for its pharmacological activity as a KSP inhibitor
comparable product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service