All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H7NO2S
CAS Number:
Molecular Weight:
169.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
98%
mp
251-255 °C (dec.) (lit.)
SMILES string
OC(=O)CSc1ccncc1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service