Skip to Content
Merck
All Photos(1)

Key Documents

182826

Sigma-Aldrich

Polyvinyltoluene, mixed isomers

average Mw ~80,000 by GPC, granular

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002

form

granular

mol wt

average Mw ~80,000 by GPC

bp

100 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C=C)cc1.Cc2cccc(C=C)c2.Cc3ccccc3C=C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service