Skip to Content
Merck
All Photos(1)

Key Documents

181358

Sigma-Aldrich

Poly(benzyl methacrylate)

average Mw ~100,000 by GPC, powder

Synonym(s):

PBMA

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$232.00

$232.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$232.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$232.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

mol wt

average Mw ~100,000 by GPC

refractive index

n20/D 1.568 (lit.)

transition temp

Tg 54 °C

density

1.179 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCc1ccccc1

InChI

1S/C11H12O2/c1-9(2)11(12)13-8-10-6-4-3-5-7-10/h3-7H,1,8H2,2H3

InChI key

AOJOEFVRHOZDFN-UHFFFAOYSA-N

Application

A variety of morphologies can be obtained by preparing poly(benzyl methacrylate)(PBzMA)/epoxy thermoset polymer blends of different compositions. The changing morphology is known to have an influence on the dynamic mechanical properties of the polymer blend. These thermoset blends find uses as matrix materials for composites and coatings.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic mechanical properties and morphology of poly (benzyl methacrylate)/epoxy thermoset blends.
Prolongo M G, et al.
Polym. Eng. Sci., 50(9), 1820-1830 (2010)

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service