Skip to Content
Merck
All Photos(2)

Key Documents

181099

Sigma-Aldrich

Poly(methyl vinyl ether-alt-maleic acid)

average Mw ~1,980,000 by LS, average Mn ~960,000, powder

Synonym(s):

Methoxyethylene-maleic acid copolymer, Methyl vinyl ether-maleic acid copolymer, Poly(maleic acid-methyl vinyl ether)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$180.00

$180.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$180.00

About This Item

Linear Formula:
[CH2CH(OCH3)CH(CO2H)CH(CO2H)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$180.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

mol wt

average Mn ~960,000
average Mw ~1,980,000 by LS

pH

~2.5 (5% in H2O)

SMILES string

COC=C.OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4.C3H6O/c5-3(6)1-2-4(7)8;1-3-4-2/h1-2H,(H,5,6)(H,7,8);3H,1H2,2H3/b2-1-;

InChI key

HHEHWCIYDICHCG-ODZAUARKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Detergent builder.

Features and Benefits

Dissolves readily in water, chelates alkaline earth cations but not toxic heavy metal ions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service