Skip to Content
Merck
All Photos(1)

Key Documents

18100

Sigma-Aldrich

Bromopolystyrene

extent of labeling: ~4 mmol/g Br loading

Synonym(s):

Copolymer of styrene and divinylbenzene, brominated, Polystyrene crosslinked with divinylbenzene, brominated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H4Br)]n
CAS Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~4 mmol/g Br loading
~60% substitution (brominated rings)

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heidi Cecilie Larsen Spång et al.
PloS one, 7(9), e45393-e45393 (2012-10-03)
It is known that targeting of antigen to antigen presenting cells (APC) increases immune responses. However, it is unclear if more than one APC-specific targeting unit in the antigenic molecule will increase responses. To address this issue, we have here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service