Skip to Content
Merck
All Photos(1)

Key Documents

180882

Sigma-Aldrich

Poly(acrylonitrile-co-butadiene-co-styrene)

acrylonitrile ~40 wt. %, powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CN)]x(CH2CH=CHCH2)y[CH2CH(C6H5)]Z
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

powder

composition

acrylonitrile, ~40 wt. %

solubility

acetone, MEK and DMF: soluble
water and alcohols: insoluble

density

1.04 g/mL at 25 °C

SMILES string

C=CC=C.C=CC#N.C=Cc1ccccc1

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

High butadiene content.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brad M Isaacson et al.
Journal of rehabilitation research and development, 46(9), 1109-1120 (2009-01-01)
Developing noninvasive tools that determine implant attachment strength to bone and monitor implant stability over time will be important to optimize rehabilitation protocols following insertion of osseointegrated implants in patients with limb loss. While resonance frequency has been previously shown
Woo-Hyuk Jung et al.
Environmental science & technology, 43(7), 2563-2568 (2009-05-21)
Cellular phone housings were ground to make original particulates using a knife mill. Foams and adhesives with a lighter density than water were removed from ground mixtures using a sink-float process in water; ground metals, button rubbers, and wires were
Bo Lai et al.
Huan jing ke xue= Huanjing kexue, 32(4), 1055-1059 (2011-07-02)
Wastewater from the condensation and drying section of acrylonitrile-butadiene-styrene (ABS) resin plant was pretreated by the microelectrolysis, and the effect of the influent pH value on the pollution removal efficiency of the microelectrolysis was mainly studied. In order to study
Martin Schlummer et al.
Waste management & research : the journal of the International Solid Wastes and Public Cleansing Association, ISWA, 24(6), 573-583 (2007-01-27)
Shredder residues produced in plants processing waste electric and electronic equipment are excluded from material recycling due to a variety of polymeric materials and the presence of brominated flame retardants (BFR), which might contain banned polybrominated diphenyl ethers or toxic
P Jyotishkumar et al.
The journal of physical chemistry. B, 113(16), 5418-5430 (2009-04-01)
The epoxy system containing diglycidyl ether of bisphenol A and 4,4'-diaminodiphenyl sulfone is modified with poly(acrylonitrile-butadiene-styrene) (ABS) to explore the effects of the ABS content on the phase morphology, mechanism of phase separation, and viscoelastic properties. The amount of ABS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service