Skip to Content
Merck
All Photos(3)

Key Documents

180262

Sigma-Aldrich

2-Methyl-2-nitrosopropane dimer

Synonym(s):

1,1,1′,1′-Tetramethylazoethane dioxide, NSC 677505, Nitroso-t-butane, Nitroso-tert-butane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$230.00
5 G
$639.00

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$230.00
5 G
$639.00

About This Item

Linear Formula:
[(CH3)3CNO]2
CAS Number:
Molecular Weight:
174.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

mp

81-83 °C (lit.)

functional group

C-nitroso
amine
nitroso

storage temp.

−20°C

SMILES string

CC(C)(C)N=O.CC(C)(C)N=O

InChI

1S/2C4H9NO/c2*1-4(2,3)5-6/h2*1-3H3

InChI key

PKKJRWITDTTZCL-UHFFFAOYSA-N

Application

2-Methyl-2-nitrosopropane dimer (Nitroso-tert-butane) was used as a spin trapping reagent in electron spin resonance studies[1].
Spin trap reagent

Reactant involved in the synthesis of air-stable π-radical based on Corannulene

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inorganic Chemistry, 31, 2333-2333 (1992)
H Sun et al.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan, 14(1), 51-55 (1994-03-01)
The free radical mechanism of enhancing radiosensibility by the Synergic Recipe to Strengthen Body Resistance (SRSBR) consisting of 10 Chinese drugs was studied by electron spin resonance (ESR) with spin trapping reagent--Nitroso-tert-butane (NtB) following irradiation of the Deoxy thymidine (dT)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service