Skip to Content
Merck
All Photos(1)

Key Documents

179078

Sigma-Aldrich

trans-2-Methylcyclopentanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C5H8OH
CAS Number:
Molecular Weight:
100.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.45 (lit.)

bp

150-151 °C/740 mmHg (lit.)

density

0.92 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@@H]1CCC[C@H]1O

InChI

1S/C6H12O/c1-5-3-2-4-6(5)7/h5-7H,2-4H2,1H3/t5-,6-/m1/s1

InChI key

BVIJQMCYYASIFP-PHDIDXHHSA-N

General description

trans-2-Methylcyclopentanol is one of the constituents of Gynandropsis gynandra oil.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gynandropsis gynandra essential oil and its constituents as tick (Rhipicephalus appendiculatus) repellents.
Phytochemistry, 50(3), 401-405 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service