Skip to Content
Merck
All Photos(1)

Key Documents

179043

Sigma-Aldrich

Borane N,N-diethylaniline complex

Synonym(s):

(N,N-Diethylaniline)trihydroboron, Diethylphenylamine-borane, NSC 239123

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$122.00
100 G
$274.00

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$122.00
100 G
$274.00

About This Item

Linear Formula:
C6H5N(C2H5)2·BH3
CAS Number:
Molecular Weight:
163.07
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reagent type: reductant

mp

−30-−27 °C (lit.)

density

0.917 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

B.CCN(CC)c1ccccc1

InChI

1S/C10H15N.BH3/c1-3-11(4-2)10-8-6-5-7-9-10;/h5-9H,3-4H2,1-2H3;1H3

InChI key

YPWBYWNNJVSNPQ-UHFFFAOYSA-N

Application

Reactant for:
  • Diastereoselective reduction of prochiral enone intermediates
  • Borylation of aryl halides
  • Reductions of ketones, acids, esters, amides, and nitriles
  • Investigations of borane source on enantioselectivity in enantiopure oxazaborolidine-catalyzed asymmetric borane reduction of ketones

Reactant for synthesis of:
  • Chromanol derivatives as CETP inhibitors for the treatment of cardiovascular disease
  • Allylic alcohols via enantioselective reductions
  • Pinacolboranes for one-pot synthesis of unsymmetrical biaryls

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service