Skip to Content
Merck
All Photos(2)

Key Documents

177962

Sigma-Aldrich

Undecanedioic acid

97%

Synonym(s):

1,9-Nonanedicarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$429.00

$429.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$429.00

About This Item

Linear Formula:
HOOC(CH2)9COOH
CAS Number:
Molecular Weight:
216.27
Beilstein:
1780537
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$429.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

108-110 °C (lit.)

SMILES string

OC(=O)CCCCCCCCCC(O)=O

InChI

1S/C11H20O4/c12-10(13)8-6-4-2-1-3-5-7-9-11(14)15/h1-9H2,(H,12,13)(H,14,15)

InChI key

LWBHHRRTOZQPDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Matsumoto et al.
Biomedical & environmental mass spectrometry, 19(3), 171-175 (1990-03-01)
Urine from patients with calcium-4-(2,4-dihydroxy-3,3-dimethylbutyramido) butyrate hemihydrate (hopantenate) therapy during episodes of Reye's-like syndrome was found to contain a number of unusual dicarboxylic acids in high concentrations; odd- and even-numbered medium-chain dicarboxylic acids, alpha-hydroxydicarboxylic acids and beta-hydroxydicarboxylic acids. The abnormal
M Claire et al.
Connective tissue research, 4(2), 61-71 (1976-01-01)
Macroscopically lesion-free parts of human aortas with no or light lesions (group I) and advanced atherosclerotic lesions (group II) were submitted to a series of successive extractions in order to "solubilize" all the macromolecular components of the arterial wall ("chemical
Shuang Yang et al.
Journal of agricultural and food chemistry, 64(48), 9237-9245 (2016-12-10)
The current study aimed to investigate whether quercetin plays a protective role in acrylamide (AA)-induced toxicity using a metabolomics approach. Rats were randomly divided into groups as follows: control, treated with AA [5 mg/kg body weight (bw)], treated with different
Xiaoling Zang et al.
Journal of proteome research, 13(7), 3444-3454 (2014-06-13)
Prostate cancer (PCa) is the second leading cause of cancer-related mortality in men. The prevalent diagnosis method is based on the serum prostate-specific antigen (PSA) screening test, which suffers from low specificity, overdiagnosis, and overtreatment. In this work, untargeted metabolomic
Fei Li et al.
Journal of proteome research, 14(4), 1937-1946 (2015-02-26)
Colon cancer is the most common cancer and the third leading cause of cancer mortality in humans. Using mass spectrometry-based metabolomics, the current study revealed the accumulation of four uremic toxins (cresol sulfate, cresol glucuronide, indoxyl sulfate, and phenyl sulfate)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service