Skip to Content
Merck
All Photos(1)

Documents

177180

Sigma-Aldrich

Methoxyacetone

95%

Synonym(s):

Methoxy-2-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2OCH3
CAS Number:
Molecular Weight:
88.11
Beilstein:
1560175
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.397 (lit.)

bp

118 °C (lit.)

density

0.957 g/mL at 25 °C (lit.)

SMILES string

COCC(C)=O

InChI

1S/C4H8O2/c1-4(5)3-6-2/h3H2,1-2H3

InChI key

CUZLJOLBIRPEFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The gas-phase basicity of methoxyacetone was studied.

Application

Methoxyacetone was used in the synthesis of doubly borylated enolates.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsushi Abiko et al.
Journal of the American Chemical Society, 124(36), 10759-10764 (2002-09-05)
The novel doubly borylated enolate is identified as an intermediate of the double aldol reaction of acetate esters. As a precursor to the formation of the doubly borylated enolate, carbon-bound boron enolates of carboxylic esters are spectroscopically characterized for the
Guy Bouchoux et al.
The journal of physical chemistry. A, 109(51), 11851-11859 (2005-12-22)
The gas-phase basicities of a representative set of hydroxy- and methoxycarbonyl compounds (hydroxyacetone, 1, 3-hydroxybutanone, 2, 3-hydroxy-3-methylbutanone, 3, 1-hydroxy-2-butanone, 4, 4-hydroxy-2-butanone, 5, 5-hydroxy-2-pentanone, 6, methoxyacetone, 7, 3-methoxy-2-butanone, 8, 4-methoxy-2-butanone, 9, and 5-methoxy-2-pentanone, 10) were experimentally determined by the equilibrium method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service