Skip to Content
Merck
All Photos(1)

Key Documents

17636

Sigma-Aldrich

5-Bromo-2-methylpyridine

≥99.0% (HPLC)

Synonym(s):

5-Bromo-2-picoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrN
CAS Number:
Molecular Weight:
172.02
Beilstein:
107324
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99.0% (HPLC)

form

solid

impurities

~5% water

mp

32-36 °C

SMILES string

Cc1ccc(Br)cn1

InChI

1S/C6H6BrN/c1-5-2-3-6(7)4-8-5/h2-4H,1H3

InChI key

OFKWIQJLYCKDNY-UHFFFAOYSA-N

Application

5-Bromo-2-methylpyridine was used in the synthesis of 6,6′-dimethyl-3,3′-bipyridine[1].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functionalised 3, 3′-bipyridines?a new class of dinucleating ligands.
Constable EC, et al.
New. J. Chem., 22(3), 287-294 (1998)
Qiuju Yin et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 204, 88-98 (2018-06-17)
In order to improve the metabolism of pirfenidone (5-methyl-1-phenylpyridin-2-one, PFD), the methyl-deuterated version of pirfenidone via the substitution of hydrogen (H) at C-5 by its isotope deuterium (D, 5D-PFD) was synthesized and its photophysical properties were investigated. The negative solvatochrom

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service