Skip to Content
Merck
All Photos(2)

Documents

175161

Sigma-Aldrich

6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione

98%

Synonym(s):

2,2-Dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CF2COCH2COC(CH3)3
CAS Number:
Molecular Weight:
296.18
Beilstein:
2056991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

impurities

0.1% 4,4′-methylenebis(2,6-di-tert-butylphenol)

refractive index

n20/D 1.379 (lit.)

bp

46-47 °C/5 mmHg (lit.)

density

1.273 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C(=O)CC(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C10H11F7O2/c1-7(2,3)5(18)4-6(19)8(11,12)9(13,14)10(15,16)17/h4H2,1-3H3

InChI key

SQNZLBOJCWQLGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione (2,2-Dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedione) was used as a co-inducer in the production of human γ interferon (HuIFN-γ). It was also used as a beta-diketone chelating agent in the extraction of supercritical carbon dioxide.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Chan Wu et al.
Journal of hazardous materials, 163(2-3), 1239-1245 (2008-09-06)
This study presents supercritical carbon dioxide (scCO(2)) extraction as an inherently safer and cleaner sample treatment method for identifying trace gallium in urine samples. Extraction is performed in the presence of a fluorinated beta-diketones chelating agent, 2,2-dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedione (HFOD), by unmodified
M Deleers et al.
Chemistry and physics of lipids, 33(1), 11-20 (1983-07-01)
The beta-diketone 1,1,1,2,2,3,3-heptafluoro-7,7-dimethyloctane-4,6-dione (FOD) translocates calcium from an aqueous medium into an organic phase. FOD is less efficient than but acts synergistically with A23187 in causing calcium translocation. The FOD-mediated process of calcium translocation is inhibited by NaCl, although the
A Kreutzberger et al.
Arzneimittel-Forschung, 31(2), 288-290 (1981-01-01)
Since in the compound class of 2-ureidopyrimidines accessible through condensation of guanidinecarboxamide (1) with beta-diketones, representatives occur possessing hypnotic activity, the range of applicability of this synthesis on the one hand and the properties of the end products as related
B D Gomperts et al.
European journal of biochemistry, 117(3), 559-562 (1981-07-01)
We show that the relatively simple beta-diketone, 1.1.1.2.2.3.3-heptafluoro-7,7-dimethyloctane-4,6-dione, known as "fod", will solvate Ca2+ ions in an organic phase. In consequence it will transport Ca2+ across phospholipid bilayers. It acts as predicted for a Ca2+-transporting ionophore in two biological systems:
I A Braude
Journal of immunological methods, 63(2), 237-246 (1983-10-14)
A method for the production of human gamma interferon (HuIFN-gamma), which is both simple and efficient, has been developed. The 2 co-inducers, A-23187 and mezerein, stimulate human peripheral blood lymphocytes to produce high levels of HuIFN-gamma in both stationary cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service