Skip to Content
Merck
All Photos(1)

Key Documents

17457

Sigma-Aldrich

Methyl α-bromoisobutyrate

≥99.0% (GC)

Synonym(s):

Methyl 2-bromo-2-methylpropionate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$70.20

$70.20


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$70.20

About This Item

Linear Formula:
(CH3)2CBrCOOCH3
CAS Number:
Molecular Weight:
181.03
Beilstein:
1209484
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$70.20


Estimated to ship onMay 26, 2025


Request a Bulk Order

Assay

≥99.0% (GC)

form

liquid

density

1.399 g/mL at 20 °C (lit.)

functional group

bromo
ester

SMILES string

COC(=O)C(C)(C)Br

InChI

1S/C5H9BrO2/c1-5(2,6)4(7)8-3/h1-3H3

InChI key

PQUSVJVVRXWKDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with a methyl end group
Methyl α-bromoisobutyrate was used in Reformatsky reaction to prepare 1,4-disubstituted 3,3-dimethylazetidin-2-ones[1].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reformatsky reaction of methyl ?-bromoisobutyrate with Schiff bases derived from salicylaldehyde and 2-hydroxynaphthalene-1-carbaldehyde.
Russ. J. Org. Chem., 40(6), 834-836 (2004)

Protocols

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service