Skip to Content
Merck
All Photos(1)

Documents

174092

Sigma-Aldrich

2-Pentyl acetate

99%

Synonym(s):

1-Methylbutyl acetate, 2-Pentyl acetat, sec.-Pentyl acetat

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH(CH3)CH2CH2CH3
CAS Number:
Molecular Weight:
130.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

availability

available only in EU

SMILES string

O=C(C)OC(CCC)C

InChI

1S/C7H14O2/c1-4-5-6(2)9-7(3)8/h6H,4-5H2,1-3H3

InChI key

GQKZRWSUJHVIPE-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.1 °F - closed cup

Flash Point(C)

31.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service