Skip to Content
Merck
All Photos(2)

Key Documents

172006

Sigma-Aldrich

Benzoin ethyl ether

97%

Synonym(s):

α-Ethoxy-α-phenylacetophenone, 2-Ethoxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$126.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$126.00

About This Item

Linear Formula:
C6H5COCH(OC2H5)C6H5
CAS Number:
Molecular Weight:
240.30
Beilstein:
2053926
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

59-61 °C (lit.)

SMILES string

CCOC(C(=O)c1ccccc1)c2ccccc2

InChI

1S/C16H16O2/c1-2-18-16(14-11-7-4-8-12-14)15(17)13-9-5-3-6-10-13/h3-12,16H,2H2,1H3

InChI key

KMNCBSZOIQAUFX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geraint J Parfitt et al.
PloS one, 7(12), e53245-e53245 (2013-01-10)
Current immunofluorescence protocols are limited as they do not provide reliable antibody staining within large tissue volumes (mm(3)) and cannot localise and quantify multiple antigens or cell populations in the same tissue at high resolution. To address this limitation, we
Katarzyna Marciniak et al.
International journal of molecular sciences, 21(5) (2020-03-12)
Precise control of generative organ development is of great importance for the productivity of crop plants, including legumes. Gibberellins (GAs) play a key role in the regulation of flowering, and fruit setting and development. The major repressors of GA signaling
Abdelmoneim Mars et al.
Mikrochimica acta, 188(3), 94-94 (2021-02-22)
A molecularly imprinted paper-based analytical device (MIP-μPAD) was developed for the sensing of bisphenol A (BPA). The platform was screen-printed onto a filter paper support, where the electrodes and the fluorescence μPADs were designed. Owing to its dual electrochemical and
W E Medina-Ortiz et al.
The open ophthalmology journal, 1, 8-16 (2007-01-01)
We have determined the presence and cellular distribution of intracellular calcium channels, inositol 1, 4, 5-trisphosphate receptors (IP3Rs) and ryanodine receptors (RyRs) in adult and postnatal (P10) lacrimal gland acinar cells. Western blot analysis of both P10 cultures and adult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service