Skip to Content
Merck
All Photos(3)

Key Documents

171719

Sigma-Aldrich

1,2-Dicyanobenzene

98%

Synonym(s):

Phthalonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$59.20
500 G
$154.00

$59.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$59.20
500 G
$154.00

About This Item

Linear Formula:
C6H4(CN)2
CAS Number:
Molecular Weight:
128.13
Beilstein:
775028
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

137-139 °C (lit.)

functional group

nitrile

SMILES string

N#Cc1ccccc1C#N

InChI

1S/C8H4N2/c9-5-7-3-1-2-4-8(7)6-10/h1-4H

InChI key

XQZYPMVTSDWCCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Dicyanobenzene was used in the synthesis of a six-coordinate phthalocyanine analogue[1].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takamitsu Fukuda et al.
Dalton transactions (Cambridge, England : 2003), 42(47), 16486-16489 (2013-10-18)
A reaction of 1,2-dicyanobenzene and lithium methoxide at 70 °C in methanol yielded the half-phthalocyanine intermediate, which coordinates to a cadmium(II) ion in the subsequent reaction step to give the first example of a six-coordinate phthalocyanine analogue containing a divalent
Juan Huang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 29(1), 207-210 (2009-04-24)
In decades, metallo-phthalocyanines (MPcs) have undergone a renaissance because of their singular and unconventional physical properties. However, for the successful application of MPcs in practical devices, it is important to disperse MPc molecules into solid state matrix to fabricate MPc
An estimation of the sublimation rate of o-phthalodinitrile and acrylamide during filter sampling.
S Kanno
Industrial health, 23(2), 167-170 (1985-01-01)
A P Kumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 53A(12), 2033-2039 (1998-01-23)
A zero-order normal coordinate analysis was made for p-, m- and o-dicyanobenzenes and their deuterated isomers by transferring the force constants from Part I. The observed and calculated frequencies agree with an average error of 18.7 cm-1, demonstrating the transferability
Xingshu Li et al.
Theranostics, 9(22), 6412-6423 (2019-10-08)
Targeted delivery of therapeutic agents is of particular interest in the field of cancer treatment. However, there is an urgent need for developing clinically promising targeting approaches that can be readily administered in a green manner. Methods: Five phthalocyanine derivatives

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service