Skip to Content
Merck
All Photos(1)

Key Documents

17049

Sigma-Aldrich

4-(Hydroxymethyl)benzaldehyde dimethyl acetal

≥97.0% (GC)

Synonym(s):

4-(Dimethoxymethyl)benzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
Beilstein:
2577568
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

solid

mp

38-42 °C

storage temp.

2-8°C

SMILES string

COC(OC)c1ccc(CO)cc1

InChI

1S/C10H14O3/c1-12-10(13-2)9-5-3-8(7-11)4-6-9/h3-6,10-11H,7H2,1-2H3

InChI key

QUWDMLYTJRQRRO-UHFFFAOYSA-N

Application

4-(Hydroxymethyl)benzaldehyde dimethyl acetal was used in the synthesis of 5′-O-TBDPS-2′,3′-O-(4-hydroxymethyl-(1R)-benzyliden)-inosine.

Other Notes

Linker employed for the immobilization of carbohydrates by acetalization and reacting the benzylic alcohol with a polymer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A solid-phase approach to the synthesis of N-1-alkyl analogues of cyclic inosine-diphosphate-ribose (cIDPR).
Tetrahedron, 66(10), 1931-1936 (2010)
T.F.J. Lampe et al.
Angewandte Chemie (International Edition in English), 110, 1761-1761 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service