Skip to Content
Merck
All Photos(1)

Key Documents

167592

Sigma-Aldrich

1-Chloro-2-methyl-2-propanol

97%

Synonym(s):

Chloro-tert-butyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$225.00

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$225.00

About This Item

Linear Formula:
(CH3)2C(OH)CH2Cl
CAS Number:
Molecular Weight:
108.57
Beilstein:
1697054
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.439 (lit.)

bp

124-128 °C (lit.)

density

1.058 g/mL at 25 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

CC(C)(O)CCl

InChI

1S/C4H9ClO/c1-4(2,6)3-5/h6H,3H2,1-2H3

InChI key

JNOZGFXJZQXOSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Reaction of 1-chloro-2-methyl-2-propanol on oxygen-containing Ag(110) surfaces has been investigated by synchrotron-based temperature-programmed X-ray photoelectron spectroscopy[1].

Application

1-Chloro-2-methyl-2-propanol was used in the synthesis and characterization of new polyfunctional thiols[2].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multistep reaction processes in epoxide formation from 1-chloro-2-methyl-2-propanol on Ag (110) revealed by TPXPS and TPD experiments.
Piao H, et al.
The Journal of Physical Chemistry B, 107(50), 13976-13985 (2003)
Catherine Vermeulen et al.
Combinatorial chemistry & high throughput screening, 9(8), 583-590 (2006-10-05)
Combinatorial chemistry was shown to be an efficient tool for the preparation of new aroma-impact compounds. In this case, polyfunctional thiols were synthesized quickly using halide reagents or Bunte salt intermediates. They were separated by gas chromatography and then characterized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service