Skip to Content
Merck
All Photos(1)

Key Documents

166391

Sigma-Aldrich

1-Formylpyrrolidine

97%

Synonym(s):

Pyrrolidine-1-carboxaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$331.00

$331.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$331.00

About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
Beilstein:
106540
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$331.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.479 (lit.)

bp

92-94 °C/15 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)N1CCCC1

InChI

1S/C5H9NO/c7-5-6-3-1-2-4-6/h5H,1-4H2

InChI key

AGRIQBHIKABLPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Formylpyrrolidine is the monomer constituent of gas clathrate inhibitor[1].

Application

1-Formylpyrrolidine was used in the synthesis of 1-oxa-3,4-dimethyl-5-(1-pyrrolldino)-2,2-di(tert-butyl)silacyclopentane and 1-oxa-4-isopropyl-5-(1-pyrrolidino)-2,2-di(tert-butyl)silacyclopentane[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and synthetic utility of oxasilacyclopentane acetals derived from siliranes.
Shaw JT and Woerpel KA.
Tetrahedron, 53(48), 16597-16606 (1997)
Computational studies of structure and dynamics of clathrate inhibitor monomers in solution.
Gomez G, et al.
Industrial & Engineering Chemistry Research, 46(1), 131-142 (2007)
T Yoshimoto et al.
Journal of biochemistry, 98(4), 975-979 (1985-10-01)
The inhibitory effects of proline-containing peptides and their derivatives on prolyl endopeptidases from Flavobacterium meningosepticum and bovine brain were compared. Replacement of the carboxyl terminal proline in N-blocked peptides with prolinal resulted in remarkable decreases in Ki values for both
Joseph Bejjani et al.
The Journal of organic chemistry, 68(25), 9747-9752 (2003-12-06)
N-Tritylprolinal (prepared in four steps from l-proline) shows a very high Felkin diastereoselectivity in its reaction with various nucleophiles, leading to a straightforward and highly stereoselective access to syn-proline-derived amino alcohols.
M Saito et al.
Journal of enzyme inhibition, 3(3), 163-178 (1990-01-01)
Several prolinal derivatives were synthesized and examined for their inhibitory activity on post-proline cleaving enzymes from Flavobacterium meningosepticum and bovine brain and their possible properties as nootropic agents. Almost all the compounds tested inhibited the activity of both enzymes at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service